L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%
CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR352680050
Price (MYR)
396.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
396.00
Add to cart
Chemical Identifiers
CAS61072-56-8
IUPAC Name4-chloro-2-fluorobenzaldehyde
Molecular FormulaC7H4ClFO
InChI KeyUVGYSEIWAOOIJR-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow to light beige
GC>=98.5 %
Appearance (Form)Powder or crystals
Melting point57°C to 61°C