trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
Acros Organics
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
CAS: 24680-50-0 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR351710250
Price (MYR)
184.42
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
184.42
Add to cart
Chemical Identifiers
CAS527-72-0
IUPAC Namethiophene-2-carboxylic acid
Molecular FormulaC5H4O2S
InChI KeyQERYCTSHXKAMIS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point126°C to 130°C
Titration with NaOH98.5 to 101.5 %
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to cream to light brown