trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
Acros Organics
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
CAS: 24680-50-0 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR351710050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-4-Methoxycinnamaldehyde
CAS24680-50-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/