Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
CAS: 80-15-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR349960050
Price (MYR)
427.00
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS9005-67-8
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
InChI KeyDEKFZWMENCCOFU-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD01780110
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid or gel or semi-solid or paste
Hydroxyl Value81.0-96.0 mgKOH/g
Acid Value≤ 2.2 mgKOH/g
Appearance (Color)Pale yellow to yellow to pale brown
Water Content (Karl Fischer Titration)≤ 3.0%
View more
Oxidation reagent
RUO – Research Use Only