Celite™ 545, Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2.5 kg
Packaging:
View all
Fiber Drum
Catalog number ACR349670100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Packaging:
Fiber Drum
Request bulk or custom format
Chemical Identifiers
CAS39637-99-5
IUPAC Name(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular FormulaC10H8ClF3O2
InChI KeyPAORVUMOXXAMPL-VIFPVBQESA-N
SMILESCO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Specific optical rotation-128° to -134° (20°C, 589 nm) (c=1, CHCl3)
Appearance (Color)Clear colorless to yellow
Refractive index1.4675 to 1.4715 (20°C, 589 nm)
View more
Filter aid
RUO – Research Use Only