Celite™ 545, Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
10 kg
Packaging:
View all
Plastic Drum
Catalog number ACR349670025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS4411-25-0
IUPAC Name1-isocyanatoadamantane
Molecular FormulaC11H15NO
InChI KeyVBHCPGFCIQDXGZ-UHFFFAOYSA-N
SMILESO=C=NC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point140°C to 150°C
Appearance (Color)White
GC>=97.5 %
Filter aid
RUO – Research Use Only