1-Aminobenzotriazole, 98%, Thermo Scientific Chemicals
1-Aminobenzotriazole, 98%, Thermo Scientific Chemicals
1-Aminobenzotriazole, 98%, Thermo Scientific Chemicals
1-Aminobenzotriazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Aminobenzotriazole, 98%, Thermo Scientific Chemicals
CAS: 1614-12-6 | C6H6N4 | 134.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Packaging:
Glass bottle
Catalog number ACR348642500
Price (MYR)
1,101.00
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,101.00
Add to cart
Chemical Identifiers
CAS174501-64-5
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC8H15F6N2P
InChI KeyIXQYBUDWDLYNMA-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
HPLC>=98.0 %
Water=<1.0 % (K.F.)