Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
CAS: 142-90-5 | C16H30O2 | 254.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number ACR347715000
Price (MYR)
1,024.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,024.00
Add to cart
Specifications
Chemical Name or MaterialLauryl methacrylate
Name Note96%
CAS142-90-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.