Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Thermo Scientific Chemicals
Flavin adenine dinucleotide disodium salt
CAS: 84366-81-4 | C27H31N9Na2O15P2 | 829.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number ACR347212500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12125-03-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
FormPowder
Assay from Supplier's CofATin (Sn): ≥37.0%