Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Thermo Scientific Chemicals
Flavin adenine dinucleotide disodium salt
CAS: 84366-81-4 | C27H31N9Na2O15P2 | 829.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number ACR347210010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFlavin adenine dinucleotide disodium salt
CAS84366-81-4
ColorYellow to Orange
EINECS Number282-733-8
Infrared SpectrumAuthentic