Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR346760250
Price (MYR)
581.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
581.00
Add to cart
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Refractive index1.5230 to 1.5250 (20°C, 589 nm)