Hemin, 99%, porcine
Hemin, 99%, porcine
Hemin, 99%, porcine
Thermo Scientific Chemicals
Hemin, 99%, porcine
Hemin, 98%, porcine, CAS # 16009-13-5, is an oxidized version of heme. It is an iron-containing prosthetic group in several proteins, and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR345960250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-65-2
IUPAC Namelithium(1+) hydroxide
Molecular FormulaHLiO
InChI KeyWMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES[Li+].[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Titration with HCl>=97.5 %
General Description
• Hemin derived from porcine is an iron-containing prosthetic group present in some proteins and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion
Applications
• Hemin is a protoporphyrin IX containing a ferric iron (Fe3+) ion with a coordinating chloride ligand
• It is used in cellular protection and controls biological mechanisms
• It can stimulate globulin synthesis
• It can be applied to study bacterial strains and growth conditions in the short-term colony assays and cell transfections
• It is used during the preparation of agar medium for bacteria culture
• This compound has been used in L929 and J774 murine macrophage cells in order to study conjugation, characterization and toxicity of lipophosphoglycan-polyacrylic acid conjugate for vaccination against leishmaniasis
RUO – Research Use Only
General References:
  1. Topuzogullari, M.; Cakir, Koc R., Dincer, Isoglu, S.; Bagirova, M.: Akdeste, Z.; Elcicek, S.; Oztel, O.N.; Yesilkir, Baydar, S.; Canim, Ates, S., Allahverdiyev, A.M. Conjugation, characterization and toxicity of lipophosphoglycan-polyacrylic acid conjugate for vaccination against leishmaniasis. J Biomed Sci. 2013 Jun 3;20(1):35.
  2. Lu, C. H.; Guo, W.; Qi, X. J.; Neubauer, A.; Paltiel, Y.; Willner, I. Hemin-G-quadruplex-crosslinked poly-N-isopropylacrylamide hydrogel: a catalytic matrix for the deposition of conductive polyaniline. Chem. Sci. 2015, 6 (11), 6659-6664.
  3. Preda, L.; Negrila, C.; Lazarescu, M. F.; Anastasescu, M.; Dobrescu, G.; Santos, E.; Lazarescu, V. Hemin interaction with bare and 4,4'-thio-bis-benzene-thiolate covered n-GaAs (110) electrodes. Phys. Chem. Chem. Phys. 2011, 13 (38), 17104-17114.