4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
CAS: 19692-45-6 | C11H15Cl | 182.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Glass Bottle
Catalog number ACR345490250
Price (MYR)
1,407.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,407.00
Add to cart
Chemical Identifiers
CAS28466-21-9
IUPAC Nametrimethyl-1H-pyrazol-4-amine
Molecular FormulaC6H11N3
InChI KeySSDGMKHZMNTWLS-UHFFFAOYSA-N
SMILESCN1N=C(C)C(N)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)100.0-106.0?C
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder