1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
CAS: 288-36-8 | C2H3N3 | 69.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR345480050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-99-6
IUPAC Name2-phenoxyethan-1-ol
Molecular FormulaC8H10O2
InChI KeyQCDWFXQBSFUVSP-UHFFFAOYSA-N
SMILESOCCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.5360 to 1.5400 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %