1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
CAS: 288-36-8 | C2H3N3 | 69.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR345480010
Price (MYR)
423.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
423.00
Add to cart
Chemical Identifiers
CAS1707-77-3
IUPAC Name1,2-bis(2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Molecular FormulaC12H22O6
InChI KeyODYBCPSCYHAGHA-UHFFFAOYNA-N
SMILESCC1(C)OCC(O1)C(O)C(O)C1COC(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)>96%
Optical Rotation+6 ? 1? (c=5 in chloroform)
Melting Point118-124? (with ≤3? range)
Water Content (Karl Fischer Titration)≤1%