Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
CAS: 10543-57-4 | C10H16N2O4 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number ACR344592500
Price (MYR)
1,352.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,352.00
Add to cart
Chemical Identifiers
CAS303-47-9
IUPAC Name(2S)-2-{[(3R)-5-chloro-8-hydroxy-3-methyl-1-oxo-3,4-dihydro-1H-2-benzopyran-7-yl]formamido}-3-phenylpropanoic acid
Molecular FormulaC20H18ClNO6
InChI KeyRWQKHEORZBHNRI-BMIGLBTASA-N
SMILESC[C@@H]1CC2=C(Cl)C=C(C(=O)N[C@@H](CC3=CC=CC=C3)C(O)=O)C(O)=C2C(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Appearance (Color)White to off-white
Appearance (Form)Powder