Brij™ C20, Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number ACR344295000
Price (MYR)
651.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
651.00
Add to cart
Specifications
Chemical Name or MaterialBrij™ C20
ColorWhite
Acid Value1mg KOH/g max.
CAS9004-95-9
Physical FormPellets