Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number ACR339421000
Price (MYR)
714.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
714.00
Add to cart
Chemical Identifiers
CAS557-21-1
IUPAC Namezinc cyanide
Molecular FormulaC2N2Zn
InChI KeyGTLDTDOJJJZVBW-UHFFFAOYSA-N
SMILES[Zn++].[C-]#N.[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5% by Zn EA
Elemental AnalysisZn: 54.58-56.80% (Theory 55.69%)
FormPowder
Appearance (Color)White