Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number ACR339420010
Price (MYR)
1,353.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,353.00
Add to cart
Chemical Identifiers
CAS143-07-7
IUPAC Namedodecanoic acid
Molecular FormulaC12H24O2
InChI KeyPOULHZVOKOAJMA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or flakes
GC>=98.5 %