n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
CAS: 540-10-3 | C32H64O2 | 480.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR338460050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexadecyl palmitate
CAS540-10-3
CAS Min %97.5
CAS Max %100.0
Melting Point52°C to 56°C