4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
CAS: 6160-80-1 | C16H16O9·2H2O | 388.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR337220050
Price (MYR)
1,262.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,262.00
Add to cart
Chemical Identifiers
CAS106-24-1
IUPAC Name(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-JXMROGBWSA-N
SMILESCC(C)=CCC\C(C)=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4745-1.4795 @ 20?C