4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
CAS: 6160-80-1 | C16H16O9·2H2O | 388.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR337220010
Price (MYR)
601.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
601.00
Add to cart
Chemical Identifiers
CAS611-70-1
IUPAC Name2-methyl-1-phenylpropan-1-one
Molecular FormulaC10H12O
InChI KeyBSMGLVDZZMBWQB-UHFFFAOYSA-N
SMILESCC(C)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Refractive Index1.5150-1.5200 @ 20?C
Assay (GC)≥96.0%