trans-Zeatin-riboside, 97%
trans-Zeatin-riboside, 97%
trans-Zeatin-riboside, 97%
trans-Zeatin-riboside, 97%
Thermo Scientific Chemicals
trans-Zeatin-riboside, 97%
CAS: 6025-53-2 | C15H21N5O5 | 351.36 g/mol
Have Questions?
Catalog NumberQuantity
ACR33720050050 mg
Catalog number ACR337200500
Price (MYR)
2,826.00
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (MYR)
2,826.00
Add to cart
Chemical Identifiers
CAS821-48-7
IUPAC Namebis(2-chloroethyl)azanium chloride
Molecular FormulaC4H10Cl3N
InChI KeyYMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES[Cl-].ClCC[NH2+]CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or flakes
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)212.0-220.0?C