Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
CAS: 51-03-6 | C19H30O5 | 338.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR334161000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52829-07-9
IUPAC Namebis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
Molecular FormulaC28H52N2O4
InChI KeyRWMYPXKVMUFMKS-UHFFFAOYSA-N
SMILESCC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
Assay (unspecified)>97.5%