2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR334082500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-71-7
IUPAC Nameheptanal
Molecular FormulaC7H14O
InChI KeyFXHGMKSSBGDXIY-UHFFFAOYSA-N
SMILESCCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Free acid (titration)≤2.0%
Identification (FTIR)Conforms
Refractive Index1.4100-1.4150 @ 20?C