2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number ACR334080010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Hydroxyethyl acrylate
Name NoteStabilized
Acidity4mg KOH/g max.
CAS818-61-1
CAS Min %96.0