Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 10 to 15%, C13H13IO8, CAS Number-87413-09-0, 75-09-2, dess martin, dess-martin, dess-martinperiodinane, dess-martin periodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess martin periodinane, dess-martin periodane, dess-martin reagent, triacetoxyperiodinane
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Catalog number ACR333110100
Price (MYR)
629.00
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (MYR)
629.00
Add to cart
Chemical Identifiers
CAS87413-09-0, 75-09-2
Specifications
Chemical Name or MaterialDess-Martin periodinane
CAS87413-09-0
Health Hazard 2GHS H Statement Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. May intensify fire; oxidizer.
CAS75-09-2
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention.
View more
oxidation reagent
RUO – Research Use Only