Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
CAS: 75-03-6 | C2H5I | 155.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR332125000
Price (MYR)
1,400.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,400.00
Add to cart
Chemical Identifiers
CAS7065-46-5
IUPAC Name3,3-dimethylbutanoyl chloride
Molecular FormulaC6H11ClO
InChI KeyBUTKIHRNYUEGKB-UHFFFAOYSA-N
SMILESCC(C)(C)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOOH=<1 %
Appearance (Form)Liquid
Titration RCOCl>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
Alkylation reagent
RUO – Research Use Only