Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
CAS: 75-03-6 | C2H5I | 155.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR332121000
Price (MYR)
539.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
539.00
Add to cart
Chemical Identifiers
CAS2725-81-7
IUPAC Name6-nitro-2H-chromen-2-one
Molecular FormulaC9H5NO4
InChI KeyRMERXEXZXIVNBF-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2OC(=O)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)185.0-192.0?C
Appearance (Color)White to cream or pale pink
Assay (GC)≥98.0%
Alkylation reagent
RUO – Research Use Only