2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
Acros Organics
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
CAS: 120-71-8 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR332080050
Price (MYR)
64.96
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
64.96
Add to cart
Chemical Identifiers
CAS1184-43-6
IUPAC Name2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular FormulaC40H28Cl2N12O10
InChI KeyVCESGVLABVSDRO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥96.0%
Appearance (Color)Yellow
Infrared spectrumConforms
FormPowder or crystals or crystalline powder