Toluene, 99.85%, for pesticide residue analysis
Toluene, 99.85%, for pesticide residue analysis
Toluene, 99.85%, for pesticide residue analysis
Toluene, 99.85%, for pesticide residue analysis
Thermo Scientific Chemicals
Toluene, 99.85%, for pesticide residue analysis
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3320700252.5 LGlass Bottle
Catalog number ACR332070025
Price (MYR)
865.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
865.00
Add to cart
Chemical Identifiers
CAS578-54-1
IUPAC Name2-ethylaniline
Molecular FormulaC8H11N
InChI KeyMLPVBIWIRCKMJV-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to red-brown
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5590 to 1.5610 (20°C, 589 nm)
GC>=97.5 %
For residue analysis
RUO – Research Use Only