Toluene, 99.85%, for pesticide residue analysis
Toluene, 99.85%, for pesticide residue analysis
Toluene, 99.85%, for pesticide residue analysis
Toluene, 99.85%, for pesticide residue analysis
Thermo Scientific Chemicals
Toluene, 99.85%, for pesticide residue analysis
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantity
ACR3320700101 L
Catalog number ACR332070010
Price (MYR)
513.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS142-96-1
IUPAC Name1-butoxybutane
Molecular FormulaC8H18O
InChI KeyDURPTKYDGMDSBL-UHFFFAOYSA-N
SMILESCCCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Residue after evaporation=<0.002 %
GC>=99.0 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
View more
For residue analysis
RUO – Research Use Only