Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals
Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals
Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals
Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR332031000100 mLGlass bottle
Catalog number ACR332031000
Price (MYR)
446.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
446.00
Add to cart
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
Melting Point18.0°C
Density1.2610g/mL
EINECS Number200-289-5