1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR331900050
Price (MYR)
338.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
338.00
Add to cart
Specifications
Chemical Name or Material1-Bromo-3, 5-dichlorobenzene
Name Note98%
CAS19752-55-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning