2-Decanol, 98%, Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
CAS: 1120-06-5 | C10H22O | 158.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ACR331600250
Price (MYR)
828.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
828.00
Add to cart
Chemical Identifiers
CAS1121-86-4
IUPAC Name1-fluoro-3-iodobenzene
Molecular FormulaC6H4FI
InChI KeyVSKSBSORLCDRHS-UHFFFAOYSA-N
SMILESFC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
StabilizerCopper powder
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
Refractive Index1.5825-1.5860 @ 20?C