1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
CAS: 2050-77-3 | C10H21I | 268.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
View all
Glass Bottle
Catalog number ACR330365000
Price (MYR)
3,073.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
3,073.00
Add to cart
Chemical Identifiers
CAS1314-80-3
SpecificationsSpecification SheetSpecification Sheet
Sulfide (S)70.7 to 73.6 %
Appearance (Color)Yellow to green
Phosphorus (P)27.3 to 28.4 %
Appearance (Form)Powder