1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
CAS: 2050-77-3 | C10H21I | 268.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR330361000
Price (MYR)
837.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
837.00
Add to cart
Chemical Identifiers
CAS7529-22-8
IUPAC Name4-methylmorpholin-4-ium-4-olate
Molecular FormulaC5H11NO2
InChI KeyLFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILESC[N+]1([O-])CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Solution
Strength49 to 51 %