N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
CAS: 79099-07-3 | C10H17NO3 | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR330271000
Price (MYR)
4,031.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,031.00
Add to cart
Chemical Identifiers
CAS4461-30-7
IUPAC Name2-chloroethanecarbonyl isocyanate
Molecular FormulaC3H2ClNO2
InChI KeyMOVMEFHWBOWMFU-UHFFFAOYSA-N
SMILESClCC(=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Appearance (Color)Clear yellow