N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
CAS: 79099-07-3 | C10H17NO3 | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR330270250
Price (MYR)
1,224.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,224.00
Add to cart
Specifications
Chemical Name or MaterialN-BOC-4-Piperidone
CAS79099-07-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0