N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
CAS: 79099-07-3 | C10H17NO3 | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR330270050
Price (MYR)
473.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
473.00
Add to cart
Chemical Identifiers
CAS619-14-7
IUPAC Name3-hydroxy-4-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyXLDLRRGZWIEEHT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)232.0-239.0?C