1-Hexyne, 98%, Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
CAS: 693-02-7 | C6H10 | 82.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR329900250
Price (MYR)
595.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
595.00
Add to cart
Chemical Identifiers
CAS10416-59-8
IUPAC Nametrimethylsilyl N-(trimethylsilyl)ethanimidate
Molecular FormulaC8H21NOSi2
InChI KeySIOVKLKJSOKLIF-UHFFFAOYSA-N
SMILESCC(O[Si](C)(C)C)=N[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4160 to 1.4190 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=94 %