Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
CAS: 98-82-8 | C9H12 | 120.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR329735000500 mLGlass bottle
Catalog number ACR329735000
Price (MYR)
399.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
399.00
Add to cart
Chemical Identifiers
CAS5892-11-5
IUPAC Name(1R,11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one tetrahydrate
Molecular FormulaC23H34N2O8
InChI KeyXLXHHXCMBNBDMP-BEYGIVOKSA-N
SMILESO.O.O.O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C7=C(C=C(OC)C(OC)=C7)[C@@]1(CCN2C4)[C@]6([H])[C@@]35[H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Loss on Drying6.41-10.25% (1.5 - 2.5 waters)
FormCrystals or powder or crystalline powder
Assay (unspecified)≥97.5% (dry wt. basis)