(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
Thermo Scientific Chemicals
(±)-Verapamil hydrochloride, 99+%
CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR329330050
Price (MYR)
1,840.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,840.00
Add to cart
Chemical Identifiers
CAS615-67-8
IUPAC Name2-chlorobenzene-1,4-diol
Molecular FormulaC6H5ClO2
InChI KeyAJPXTSMULZANCB-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=88 %