(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
Thermo Scientific Chemicals
(±)-Verapamil hydrochloride, 99+%
CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR329330010
Price (MYR)
640.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
640.00
Add to cart
Specifications
Chemical Name or Material(±)-Verapamil hydrochloride
CAS152-11-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0