Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
CAS: 21829-25-4 | C17H18N2O6 | 346.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR329270050
Price (MYR)
1,134.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,134.00
Add to cart
Chemical Identifiers
CAS630-93-3
IUPAC Namesodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide
Molecular FormulaC15H11N2NaO2
InChI KeyFJPYVLNWWICYDW-UHFFFAOYSA-M
SMILES[Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Color of solutionPasses test
Appearance (Form)Crystalline powder
Water=<3.0 %
Appearance (Color)White to off-white