Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
CAS: 21829-25-4 | C17H18N2O6 | 346.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Catalog number ACR329270010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤70.0%
Elemental AnalysisPd: 2.25-2.75%
Elemental AnalysisPt: 2.25-2.75%