Griess Reagent
Griess Reagent
Griess Reagent
Thermo Scientific Chemicals
Griess Reagent
Have Questions?
Catalog NumberQuantity
ACR32867050050 mL
Catalog number ACR328670500
Price (MYR)
1,121.00
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (MYR)
1,121.00
Add to cart
Chemical Identifiers
CAS93106-60-6
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Purity≥95.0%
Appearance (Color)Pale yellow
Mass SpecConforms