Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
CAS: 446-72-0 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Catalog number ACR328270250
Price (MYR)
853.00
Add to cart
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
853.00
Add to cart
Chemical Identifiers
CAS24949-35-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.5155-1.5205 @ 20?C