Fumonisin B1, 96%, Thermo Scientific Chemicals
Fumonisin B1, 96%, Thermo Scientific Chemicals
Fumonisin B1, 96%, Thermo Scientific Chemicals
Fumonisin B1, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fumonisin B1, 96%, Thermo Scientific Chemicals
Fumonisin, CAS # 116355-83-0, also known as Macrofusine, is a cell permeable mycotoxin that is an inhibitor of sphingosine N-acetyl transferase.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number ACR328250010
Price (MYR)
1,915.00
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (MYR)
1,915.00
Add to cart
Chemical Identifiers
CAS543-49-7
IUPAC Nameheptan-2-ol
Molecular FormulaC7H16O
InChI KeyCETWDUZRCINIHU-UHFFFAOYNA-N
SMILESCCCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.0 %
Specific gravity(20°C) 0.816 to 0.819
Appearance (Form)Liquid
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
View more
General Description
• Fumonisin is a fungal metabolite that is a cell permeable mycotoxin that can halt sphingolipid biosynthesis by inhibiting sphingosine N-acetyl transferase
Applications
• It can potently inhibit sphingosine N-acyltransferase (ceramide synthase), causing an accumulation of sphingoid bases
• It is also a protein serine and threonine phosphatase inhibitor
• It can selectively inhibit sphingomyelin biosynthesis in neuronal cells
RUO – Research Use Only
General References:
  1. Stockmann-Juvala, H., et al., 2008. A review of the toxic effects and mechanisms of action of fumonisin B1. Human & experimental toxicology. 27(11), 799-809.
  2. Fukuda et al (1996) Inhibition of protein serine/threonine phosphatases by Fumonisin B1, a mycotoxin. Biochem.Biophys.Res.Comms. 220 160.