Daidzein, 98%, Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
CAS: 486-66-8 | C15H10O4 | 254.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Packaging:
Glass bottle
Catalog number ACR328231000
Price (MYR)
1,844.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,844.00
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.